Learn More
Medchemexpress LLC 3-methyl-5-[(2S)-1-methyl-2-pyrrolidinyl]isoxazole hydrochloride | 147388-83-8 | MFCD11045960 | 99.4% | 202.68 g·mol⁻¹ | C9H15ClN2O | 1 ML
SDP
Catalog No. 5000243190
Encompass_Preferred
ABT-418 hydrochloride is a potent, selective agonist of neuronal nicotinic acetylcholine receptors (nAChRs) used in preclinical research to investigate cholinergic signaling, cognition, and anxiolytic mechanisms. It is supplied as a solid and as a ready-to-use 10 mM solution for laboratory applications. CAS 147388-83-8; molecular weight 202.68 g·mol⁻¹. For research use only.
  • Potent, selective agonist of neuronal nicotinic acetylcholine receptors.
  • Available as solid and as 10 mM solution ready for use.
  • High purity suitable for analytical and biological studies.
  • Stable under recommended storage conditions.
  • Intended for laboratory research use only.
Catalog No. 50-002-43190 Supplier Medchemexpress LLC Supplier No. HY105170B10MM 1ML
ABT-418 hydrochloride is a potent, selective agonist of neuronal nicotinic acetylcholine receptors (nAChRs) used in preclinical research to investigate cholinergic signaling, cognition, and anxiolytic mechanisms. It is supplied as a solid and as a ready-to-use 10 mM solution for laboratory applications. CAS 147388-83-8; molecular weight 202.68 g·mol⁻¹. For research use only.
  • Potent, selective agonist of neuronal nicotinic acetylcholine receptors.
  • Available as solid and as 10 mM solution ready for use.
  • High purity suitable for analytical and biological studies.
  • Stable under recommended storage conditions.
  • Intended for laboratory research use only.