Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-5-(trifluoromethyl)benzonitrile, 97%

Catalog No. AAH3227109
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3227109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3227109 Supplier Thermo Scientific Chemicals Supplier No. H3227109
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 261952-04-9 | C9H6F3N | 185.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261952-04-9
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD01631595
InChI Key CSJHJNBBWUGJFY-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethyl benzonitrile, benzonitrile,3-methyl-5-trifluoromethyl, 3-cyano-5-methylbenzotrifluoride, 3-methyl-5-trifluoromethyl-benzonitrile, 3-methyl-5-trifluoromethyl benzenecarbonitrile, 5-methyl-3-trifluoromethyl benzenecarbonitrile
PubChem CID 2775595
IUPAC Name 3-methyl-5-(trifluoromethyl)benzonitrile
SMILES CC1=CC(=CC(=C1)C#N)C(F)(F)F

Specifications

Refractive Index 1.463
Quantity 10 g
UN Number UN3276
Formula Weight 185.15
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)benzonitrile
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.