Learn More
Medchemexpress LLC 3-methyl hippuric acid-d7 | 1216551-07-3 | 99.0% | 200.24 g/mol | C10H4D7NO3 | 5 MG
SDP
Catalog No. 5000263040
Encompass_Preferred
3-Methyl Hippuric acid-d7 is a deuterium-labeled analogue of 3-methyl hippuric acid used as an internal standard for quantitative mass spectrometry workflows. Supplied as a white to off-white solid with high isotopic purity, it is intended for accurate, reproducible quantification of 3-methyl hippuric acid in biological samples such as urine and plasma.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • Improves quantification accuracy and precision.
  • Suitable for urine, plasma, and serum sample analysis.
  • High chemical and isotopic purity for reliable results.
  • Supplied as a stable, white to off-white solid.
  • Easy to prepare and spike into calibration standards and controls.
Catalog No. 50-002-63040 Supplier Medchemexpress LLC Supplier No. HY131619S5MG
3-Methyl Hippuric acid-d7 is a deuterium-labeled analogue of 3-methyl hippuric acid used as an internal standard for quantitative mass spectrometry workflows. Supplied as a white to off-white solid with high isotopic purity, it is intended for accurate, reproducible quantification of 3-methyl hippuric acid in biological samples such as urine and plasma.
  • Deuterium-labeled internal standard for LC-MS/MS.
  • Improves quantification accuracy and precision.
  • Suitable for urine, plasma, and serum sample analysis.
  • High chemical and isotopic purity for reliable results.
  • Supplied as a stable, white to off-white solid.
  • Easy to prepare and spike into calibration standards and controls.