Learn More
Medchemexpress LLC 3-methyl hippuric acid-d7 | 1216551-07-3 | 99.0% | C10H4D7NO3 | 10MG
SDP
Catalog No. 5000260835
Encompass_Preferred
3-Methyl Hippuric acid-d7 is a deuterium-labeled analogue of 3-methyl hippuric acid intended as an isotopically labeled internal standard for analytical assays. It provides consistent, traceable signal response for quantitative mass spectrometry and NMR applications and is supplied as a high-purity solid for method development and calibration.
  • Deuterium-labeled internal standard for LC-MS, GC-MS, and NMR.
  • High isotopic enrichment for accurate quantitation.
  • High chemical purity suitable for analytical workflows.
  • Available in multiple small pack sizes for method development.
  • Stable solid form for convenient storage and handling.
Catalog No. 50-002-60835 Supplier Medchemexpress LLC Supplier No. HY131619S10MG
3-Methyl Hippuric acid-d7 is a deuterium-labeled analogue of 3-methyl hippuric acid intended as an isotopically labeled internal standard for analytical assays. It provides consistent, traceable signal response for quantitative mass spectrometry and NMR applications and is supplied as a high-purity solid for method development and calibration.
  • Deuterium-labeled internal standard for LC-MS, GC-MS, and NMR.
  • High isotopic enrichment for accurate quantitation.
  • High chemical purity suitable for analytical workflows.
  • Available in multiple small pack sizes for method development.
  • Stable solid form for convenient storage and handling.