Learn More
Medchemexpress LLC 3-Methyl-N-[4-(trifluoromethyl)phenyl]-2-benzofurancarboxamide | 588674-64-0 | C17H12F3NO2 | 25 MG
SDP
Catalog No. 5000374409
Encompass_Preferred
CCR6 antagonist 1 is a CCR6 antagonist that inhibits the CCL20/CCR6 axis. It is used in the research of autoimmune-mediated inflammatory diseases, such as inflammatory bowel diseases (IBDs).
  • Inhibits the CCL20/CCR6 axis
  • Blocks CCL20-induced CD4+ T cell migration
  • Alleviates TNBS-induced inflammatory responses in mice
  • Shows anti-inflammatory effects in Zymosan-induced peritonitis mice
Catalog No. 50-003-74409 Supplier Medchemexpress LLC Supplier No. 25MGHY151435
CCR6 antagonist 1 is a CCR6 antagonist that inhibits the CCL20/CCR6 axis. It is used in the research of autoimmune-mediated inflammatory diseases, such as inflammatory bowel diseases (IBDs).
  • Inhibits the CCL20/CCR6 axis
  • Blocks CCL20-induced CD4+ T cell migration
  • Alleviates TNBS-induced inflammatory responses in mice
  • Shows anti-inflammatory effects in Zymosan-induced peritonitis mice