Learn More
Medchemexpress LLC HY-19312 100mg Medchemexpress, 3-Methyladenine CAS:5142-23-4 Purity:>98%
SDP
Catalog No. 502029275
Encompass_Preferred
Medchemexpress, HY-19312 100mg 3-Methyladenine CAS:5142-23-4 3-Methyladenine (3-MA) is a PI3K inhibitor. 3-Methyladenine is a widely used inhibitor of autophagy via its inhibitory effect on class III PI3K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9275 Supplier Medchemexpress LLC Supplier No. HY19312100MG
Medchemexpress, HY-19312 100mg 3-Methyladenine CAS:5142-23-4 3-Methyladenine (3-MA) is a PI3K inhibitor. 3-Methyladenine is a widely used inhibitor of autophagy via its inhibitory effect on class III PI3K. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.