Learn More
TARGETMOL CHEMICALS INC 3-Methyladenine 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1879200MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-Methyladenine (3-MA) is a PI3K inhibitor that selectively inhibits class IB PI3Kgamma (IC50 = 60 uM) and class III VPS34 (IC50 = 25 uM). 3-Methyladenine inhibits autophagy. Purity 99.51%
Catalog No. 50-287-8335