Learn More
AdipoGen 3-Methyladenine
SDP
Supplier:  AdipoGen CDXM0203M100
Encompass_Preferred
Chemical. CAS 5142-23-4. Formula C6H7N5. MW 149.15. Synthetic Used to inhibit and study the mechanism of autophagy lysosomal self-degradation and apoptosis under various conditions. Inhibits autophagy by blocking autophagosome formation via the inhibition of type III Phosphatidylinositol 3-kinases PI-3K.
Catalog No. 50-256-0480