Learn More
AdipoGen 3-Methyladenine,Chemical. CAS: 5142-23-4
SDP
Catalog No. 501147173
Encompass_Preferred
3-Methyladenine, AdipoGen Life Sciences. Chemical. CAS: 5142-23-4. Formula: C6H7N5. MW: 149.15. Synthetic Used to inhibit and study the mechanism of autophagy (lysosomal self-degradation) and apoptosis under various conditions. Inhibits autophagy by blocking autophagosome formation via the inhibition of type III Phosphatidylinositol 3-kinases (PI-3K).
Catalog No. 50-114-7173 Supplier AdipoGen Supplier No. CDXM0203M025
3-Methyladenine, AdipoGen Life Sciences. Chemical. CAS: 5142-23-4. Formula: C6H7N5. MW: 149.15. Synthetic Used to inhibit and study the mechanism of autophagy (lysosomal self-degradation) and apoptosis under various conditions. Inhibits autophagy by blocking autophagosome formation via the inhibition of type III Phosphatidylinositol 3-kinases (PI-3K).