Learn More
Enzo Life Sciences 3-Methyladenine (25mg). CAS: 5142-23-4
SDP
Supplier:  Enzo Life Sciences BMLAP5020025
Encompass_Preferred
PI3 kinase inhibitor and a well-known inhibitor of autophagy. This compound has been shown to induce autophagy under nutrient-rich conditions as well as inhibiting starvation-induced autophagy due to differential effects on class I versus class III PI3 kinase. Alternative name: 6-Amino-3-methylpurine. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10 mg/ml) or 100% ethanol (5 mg/ml, warm). Long Term Storage: -20°C.
Catalog No. 50-201-0318