Learn More
AdipoGen 3-Methyladenine, 5142-23-4, MFCD00010531, 25 mg
SDP
Supplier:  AdipoGen AGCR13597M025
Encompass_Preferred
MF: C6H7N5, MW: 149.2, ≥98%, Appearance: White to off-white solid. Potent cell permeable and selective inhibitor of phosphatidyl-inositol 3-kinase (PI3K). Blocks class I, class II and class III PI3Ks, including some downstream targets. Induces autophagy under nutrient-rich conditions and inhibits starvation-induced autophagy due to differential effects on class I versus class III PI3 kinase
Catalog No. 50-101-6605