Learn More
Apexbio Technology LLC 3-Methyladenine, 5142-23-4, MFCD00010531, 50 mg
SDP
Catalog No. 501010207
Encompass_Preferred
MF: C6H7N5, MW: 149.15, Solubility: 7.45mg/mL in DMSO, 5mg/mL in H2O. 3-Methyladenine is an inhibitor of class III phosphoinositide 3-kinase (PI3K).
Catalog No. 50-101-0207 Supplier Apexbio Technology LLC Supplier No. A835350
MF: C6H7N5, MW: 149.15, Solubility: 7.45mg/mL in DMSO, 5mg/mL in H2O. 3-Methyladenine is an inhibitor of class III phosphoinositide 3-kinase (PI3K).