Learn More
Selleck Chemical LLC 3-Methyladenine (3-MA) 50mg 5142-23-4 NSC 66389
SDP
Supplier:  Selleck Chemical LLC S276750MG
Encompass_Preferred
3-Methyladenine (3-MA, NSC 66389) is a selective PI3K inhibitor for Vps34 and PI3K? with IC50 of 25 ?M and 60 ?M in HeLa cells; blocks class I PI3K consistently, whereas suppression of class III PI3K is transient, and also blocks autophagosome formation. 3-Methyladenine (3-MA) is successfully used to suppress mitophagy. Solutions of 3-MA are best fresh-prepared by heating. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0737715