Learn More
Medchemexpress LLC 3-methyladenine-d3 | 110953-39-4 | 99.4% | 152.17 g/mol | C6H4D3N5 | 10 MG
SDP
Catalog No. 5000294931
Encompass_Preferred
3-Methyladenine-d3 is the deuterium-labeled analogue of 3-methyladenine, provided for use as a tracer and internal standard in analytical and research applications. It supports quantitative studies related to PI3K-mediated autophagy and is suitable for mass spectrometry and NMR workflows.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable as a tracer in quantitative analytical workflows.
  • High purity (99.44%).
  • Molecular weight 152.17 g/mol and formula C6H4D3N5 for exact mass calculations.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-94931 Supplier Medchemexpress LLC Supplier No. HY19312S10MG
3-Methyladenine-d3 is the deuterium-labeled analogue of 3-methyladenine, provided for use as a tracer and internal standard in analytical and research applications. It supports quantitative studies related to PI3K-mediated autophagy and is suitable for mass spectrometry and NMR workflows.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable as a tracer in quantitative analytical workflows.
  • High purity (99.44%).
  • Molecular weight 152.17 g/mol and formula C6H4D3N5 for exact mass calculations.
  • Intended for research use only; not for human or clinical use.