Learn More
Cayman Chemical 3-MethyladIpIc AcId 25g
SDP
Supplier:  Cayman Chemical 3645225 G
Encompass_Preferred
A catabolite of phytanic acid; urinary levels have been used as a biomarker for the ω-oxidation of phytanic acid in patients with adult Refsum disease
Catalog No. 50-291-2489