Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methyladipic acid | 3058-01-3 | ≥98.0% | 160.17 | 1 G
SDP

Catalog No. 5000436970
Encompass_Preferred

3-Methyladipic acid is an endogenous metabolite and the final metabolite in the ω-oxidation pathway. This white to off-white solid, with chemical formula C7H12O4, is classified under ketones, aldehydes, and acids, and originates from microorganisms. Primarily for research use, it is stable under recommended conditions for diverse laboratory applications.

  • Endogenous metabolite
  • White to off-white solid
  • Chemical formula C7H12O4
  • Classified as ketones, aldehydes, acids
  • Originates from microorganisms
  • For research use only
  • Stable under recommended storage
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-36970 Supplier Medchemexpress LLC Supplier No. HY1132771G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methyladipic acid is an endogenous metabolite and the final metabolite in the ω-oxidation pathway. This white to off-white solid, with chemical formula C7H12O4, is classified under ketones, aldehydes, and acids, and originates from microorganisms. Primarily for research use, it is stable under recommended conditions for diverse laboratory applications.

  • Endogenous metabolite
  • White to off-white solid
  • Chemical formula C7H12O4
  • Classified as ketones, aldehydes, acids
  • Originates from microorganisms
  • For research use only
  • Stable under recommended storage
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.