Learn More
Medchemexpress LLC HY-113277 500mg Medchemexpress, 3-Methyladipic acid CAS:3058-01-3 Purity:>98%
SDP
Catalog No. 501969237
Encompass_Preferred
Medchemexpress, HY-113277 500mg 3-Methyladipic acid CAS:3058-01-3 3-Methyladipic acid is the final metabolite in the ω-oxidation pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9237 Supplier Medchemexpress LLC Supplier No. HY113277500MG
Medchemexpress, HY-113277 500mg 3-Methyladipic acid CAS:3058-01-3 3-Methyladipic acid is the final metabolite in the ω-oxidation pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.