Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methylamino-azetidine-1-carboxylic acid tert-butyl ester

Catalog No. VBP000601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VBP000601G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. VBP000601G Supplier Sigma Aldrich Supplier No. VBP000601G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Chloroacetaldehyde is a reagent used for converting adenine and its nucleoseides into fluorescent etheno derivatives which can be analysed by HPLC
  • It was used for derivatization in fluorometric determination of arprinocid and analogous compounds in human plasma .chloroacetaldehyde, a metabolite of ifosamide, influences oxidative phosphorylation in mitochondria
  • It causes breaking of DNA strands and strongly inhibits DNA synthesis.prices for bulk quantities on request

Specifications

CAS 454703-20-9
Molecular Formula C9H18N2O2
Linear Formula C9H18N2O2
MDL Number MFCD08061962
Quantity 1 kg
Molecular Weight (g/mol) 186.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.