Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylamino)pyridine 98.0+%, TCI America™
SDP

Catalog No. M26845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26845G 5 g
M26841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26845G Supplier TCI America Supplier No. M26845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18364-47-1 | C6H8N2 | 108.14 g/mol

Chemical Identifiers

CAS 18364-47-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD01418130
InChI Key DBGFNLVRAFYZBI-UHFFFAOYSA-N
Synonym n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
PubChem CID 140376
IUPAC Name N-methylpyridin-3-amine
SMILES CNC1=CC=CN=C1

Specifications

Boiling Point 110°C
Quantity 5 g
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 3-(Methylamino)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.