Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylamino)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. M11065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11065G Supplier TCI America Supplier No. M11065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 83030-08-4
Color Colorless
Boiling Point 165°C
Molecular Formula C5H12N2
Quantity 5 g
MDL Number MFCD00191460
UN Number 2734
InChI Key NGZYRKGJWYJGRS-UHFFFAOYSA-N
SMILES CNC1CCNC1
IUPAC Name N-methylpyrrolidin-3-amine
Molecular Weight (g/mol) 100.165
PubChem CID 11788268
Formula Weight 100.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Methylamino)pyrrolidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.