Learn More
Cayman Chemical 3-Methyladenine, 5142-23-4, 25 mg
SDP
Supplier:  Cayman Chemical 1324225
Encompass_Preferred
Molecular Formula C6H7N5, Purity ≥95%, Formula Weight 149.2, Synonyms: 3-methyl-3H-purin-6-amine, 3-MA, NSC 66389; A specific inhibitor of PI3K activity and a widely used inhibitor of the autophagy; at 5 mM, inhibits protein degradation in rat hepatocytes by 65% and blocks class I, II, and III PI3Ks, including some downstream targets.
Catalog No. NC1522953