Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzoic anhydride, 97%

Catalog No. p-7031607
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3273903 1 g
AAH3273906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3273903 Supplier Thermo Scientific Chemicals Supplier No. H3273903
Add to Cart

CAS: 21436-44-2 | C16H14O3 | 254.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21436-44-2
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00048083
InChI Key DGZSQLHPEZFGSI-UHFFFAOYSA-N
Synonym 3-methylbenzoic anhydride, m-toluic anhydride, 3-methylbenzene-1-carboxylic anhydride, benzoic acid, 3-methyl-, anhydride, 3-methylbenzene-1-carboxylicanhydride, 3-methylbenzoicanhydride, bis 3-methylbenzoic acid anhydride, 3-methylbenzoyl 3-methylbenzoate, benzoic acid,3-methyl-, 1,1'-anhydride
PubChem CID 253301
IUPAC Name (3-methylbenzoyl) 3-methylbenzoate
SMILES CC1=CC=CC(=C1)C(=O)OC(=O)C2=CC(=CC=C2)C

Specifications

Melting Point 66°C to 70°C
Quantity 1 g
Formula Weight 254.29
Percent Purity 97%
Chemical Name or Material 3-Methylbenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.