Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbenzothiazolium Iodide 98.0+%, TCI America™
SDP

Catalog No. M113125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M113125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M113125G Supplier TCI America Supplier No. M113125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2786-31-4
Molecular Formula C8H8INS
Molecular Weight (g/mol) 277.123
MDL Number MFCD00050297
InChI Key SGYIRNXZLWJMCR-UHFFFAOYSA-M
Synonym 3-methylbenzothiazolium iodide, 3-methyl-1,3-benzothiazol-3-ium iodide, benzothiazole methiodide, 3-methylbenzothiazoliumiodide, benzothiazolium, 3-methyl-, iodide, acmc-1cioz, benzothiazolium, 3-methyl-, iodide 1:1, n-methylbenzothiazolium iodide, benzothiazolium,3-methyl-, iodide 1:1
PubChem CID 2735081
IUPAC Name 3-methyl-1,3-benzothiazol-3-ium;iodide
SMILES C[N+]1=CSC2=CC=CC=C21.[I-]

Specifications

Quantity 25 g
Formula Weight 277.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylbenzothiazolium Iodide
EINECSNumber (5)-0231
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.