Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylbutanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018475 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8475 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8475 Supplier MilliporeSigma Supelco Supplier No. 590925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methylbutanol belongs to the group of high chain alcohols produced as metabolites from the degradation of amino acids via keto acids. It is a volatile flavor compound, highly present in alcoholic beverages.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123-51-3
Density 0.809 g/mL (at 20°C)
Physical Form Neat
Linear Formula (CH3)2 CHCH2CH2OH
MDL Number MFCD00002934
Boiling Point 131°C to 132°C
Synonym 3-Methyl-1-butanol; Isoamyl alcohol; Isopentyl alcohol
Quantity 5 mL
Formula Weight 88.15
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.