Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-methylcarbazole | 4630-20-0 | MFCD00464338 | 181.23 g/mol | C13H11N | 500 MG
SDP

Catalog No. 5000210125
Encompass_Preferred

3-methylcarbazole is a carbazole alkaloid used for research that exhibits anticancer activity. It shows growth inhibitory activity against human fibrosarcoma HT-1080 cells (IC50 = 25 μg/mL). Chemical identifiers include CAS 4630-20-0, formula C13H11N, and molecular weight 181.23 g/mol.

  • Chemical formula: C13H11N.
  • Molecular weight: 181.23 g/mol.
  • CAS number: 4630-20-0.
  • Reported bioactivity: inhibits HT-1080 cell growth (IC50 = 25 μg/mL).
  • Synonyms: NSC 10154.
  • Typical packaging: 500 mg for research use.

Catalog No. 50-002-10125 Supplier Medchemexpress LLC Supplier No. HYW047478500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-methylcarbazole is a carbazole alkaloid used for research that exhibits anticancer activity. It shows growth inhibitory activity against human fibrosarcoma HT-1080 cells (IC50 = 25 μg/mL). Chemical identifiers include CAS 4630-20-0, formula C13H11N, and molecular weight 181.23 g/mol.

  • Chemical formula: C13H11N.
  • Molecular weight: 181.23 g/mol.
  • CAS number: 4630-20-0.
  • Reported bioactivity: inhibits HT-1080 cell growth (IC50 = 25 μg/mL).
  • Synonyms: NSC 10154.
  • Typical packaging: 500 mg for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.