Learn More
Medchemexpress LLC 3-Methylcatechol | 488-17-5 | 98.2% | 124.14 | 250 MG
SDP
Catalog No. 5000451579
Encompass_Preferred
3-Methylcatechol is a phenolic derivative that can be produced by Pseudomonas putida MC2. It functions as a molecular building block in chemical synthesis for the production of other active compounds.
  • Phenolic derivative
  • Can be produced by Pseudomonas putida MC2
  • Molecular building block in chemical synthesis
Catalog No. 50-004-51579 Supplier Medchemexpress LLC Supplier No. 250MGHYW012815
3-Methylcatechol is a phenolic derivative that can be produced by Pseudomonas putida MC2. It functions as a molecular building block in chemical synthesis for the production of other active compounds.
  • Phenolic derivative
  • Can be produced by Pseudomonas putida MC2
  • Molecular building block in chemical synthesis