Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylcatechol 99.0+%, TCI America™
SDP

Catalog No. M018425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M018425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M018425G Supplier TCI America Supplier No. M018425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 488-17-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00016435
InChI Key PGSWEKYNAOWQDF-UHFFFAOYSA-N
Synonym 3-methylcatechol, 2,3-dihydroxytoluene, 3-methylpyrocatechol, 1,2-benzenediol, 3-methyl, 2,3-toluenediol, dihydroxytoluene, pyrocatechol, 3-methyl, catechol, 3-methyl, 1,2-dihydroxy-3-methylbenzene, 3-methyl-1,2-benzenediol
PubChem CID 340
ChEBI CHEBI:18404
IUPAC Name 3-methylbenzene-1,2-diol
SMILES CC1=C(C(=CC=C1)O)O

Specifications

Melting Point 66°C
Boiling Point 125°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylcatechol
EINECSNumber (4)-0057
RTECSNumber UX1910000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.