Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylcholanthrene, MilliporeSigma™ Supelco™

Catalog No. 111019742 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9742 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9742 Supplier MilliporeSigma Supelco Supplier No. 442388
Add to Cart
Add to Cart

3-Methylcholanthrene, a potent polycyclic aryl hydrocarbon receptor agonist, which is a carcinogen that can be obtained as a byproduct of combustion of various organic substances like coal, garbage, gas and oil. It suppresses the proliferation of human umbilical vascular endothelial cells.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 56-49-5
Physical Form Neat
Linear Formula C21H16
MDL Number MFCD00003704
Melting Point 178°C to 180°C
Boiling Point 280°C (80 mmHg)
Type Organic Pollutant Standard
Quantity 50 mg
Formula Weight 268.35
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.