Learn More
Cayman Chemical 3-Methylcrotonyl Glycine, Purity: ≥98%, CAS: 33008-07-0, 50 mg
SDP
Catalog No. 502993931
Encompass_Preferred
A metabolite; found in urine of patients with 3-methycrotonyl glycinuria; inhibits CO2 production and mitochondrial complex II-III and creatine kinase activity in rat cerebral cortex preparations in a concentration-dependent manner; inhibits the Na+/K+-ATPase in purified synaptic membranes from rat cerebrum
Catalog No. 50-299-3931 Supplier Cayman Chemical Supplier No. 2288850 MG
A metabolite; found in urine of patients with 3-methycrotonyl glycinuria; inhibits CO2 production and mitochondrial complex II-III and creatine kinase activity in rat cerebral cortex preparations in a concentration-dependent manner; inhibits the Na+/K+-ATPase in purified synaptic membranes from rat cerebrum