Learn More
Medchemexpress LLC 3-Methylcytidine | 2140-64-9 | 99.3% | 257.24 | 50 MG
SDP
Catalog No. 5000438878
Encompass_Preferred
3-Methylcytidine is a urinary nucleoside that can be used as a biomarker for four different types of cancer: lung cancer, gastric cancer, colon cancer, and breast cancer.
  • Utility as a biomarker for detecting lung, gastric, colon, and breast cancers.
Catalog No. 50-004-38878 Supplier Medchemexpress LLC Supplier No. HY11164550MG
3-Methylcytidine is a urinary nucleoside that can be used as a biomarker for four different types of cancer: lung cancer, gastric cancer, colon cancer, and breast cancer.
  • Utility as a biomarker for detecting lung, gastric, colon, and breast cancers.