Learn More
Medchemexpress LLC 3-methylcytidine | 2140-64-9 | MFCD07778656 | 99.3% | 257.24 g·mol⁻¹ | C10H15N3O5 | 5 MG
SDP
Catalog No. 5000263685
Encompass_Preferred
3-Methylcytidine is a modified nucleoside used as a urinary biomarker for multiple cancer types. It is provided as a solid suitable for analytical and biomarker research applications.
  • Urinary nucleoside biomarker for lung, gastric, colon, and breast cancer.
  • High purity: 99.3%.
  • Molecular formula C10H15N3O5; molecular weight 257.24 g·mol⁻¹.
  • Supplied as a solid white to off-white powder.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small research quantities such as 5 mg.
Catalog No. 50-002-63685 Supplier Medchemexpress LLC Supplier No. HY1116455MG
3-Methylcytidine is a modified nucleoside used as a urinary biomarker for multiple cancer types. It is provided as a solid suitable for analytical and biomarker research applications.
  • Urinary nucleoside biomarker for lung, gastric, colon, and breast cancer.
  • High purity: 99.3%.
  • Molecular formula C10H15N3O5; molecular weight 257.24 g·mol⁻¹.
  • Supplied as a solid white to off-white powder.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small research quantities such as 5 mg.