Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylene-2-norbornanone 97.0+%, TCI America™
SDP

Catalog No. M16565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16565G Supplier TCI America Supplier No. M16565G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5597-27-3
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
InChI Key FNOOZJAPZFHNCW-UHFFFAOYSA-N
Synonym 3-Methylenebicyclo[2.2.1]heptan-2-one
PubChem CID 96875
IUPAC Name 2-methylidenebicyclo[2.2.1]heptan-3-one
SMILES C=C1C2CCC(C2)C1=O

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 g
UN Number 1224
Formula Weight 122.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylene-2-norbornanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.