Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylfuran, 98%

Catalog No. ac342300010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC342300010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC342300010 Supplier Thermo Scientific Chemicals Supplier No. 342300010
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 930-27-8 | C5H6O | 82.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-27-8
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.1
MDL Number MFCD00060134
InChI Key KJRRQXYWFQKJIP-UHFFFAOYSA-N
Synonym furan, 3-methyl, 3-methylfurane, 3-methyl furan, 3-methyl-furan, 3-methyl-fura, 3-methylfuran, pubchem19993, 3-methylfuran-methyl-d3, acmc-20an04, 3-methylfuran 1gr
PubChem CID 13587
IUPAC Name 3-methylfuran
SMILES CC1=COC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 65.0°C to 66.0°C
Flash Point −22°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330
Quantity 1 g
Specific Gravity 0.92
Formula Weight 82.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methylfuran, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.