Learn More
Cayman Chemical 3-Methylglutaric Acid, Purity: ≥95%, CAS: 626-51-7, 10 g
SDP
Catalog No. 502949954
Encompass_Preferred
A metabolite of L-leucine; inhibits Na+/K+-ATPase activity and induces the production of ROS in rat cortical synapse preparations, as well as induces lipid peroxidation in rat cortical supernatants; urinary levels are increased in patients with 3-methylglutaconic aciduria type I
Catalog No. 50-294-9954 Supplier Cayman Chemical Supplier No. 3435710 G
A metabolite of L-leucine; inhibits Na+/K+-ATPase activity and induces the production of ROS in rat cortical synapse preparations, as well as induces lipid peroxidation in rat cortical supernatants; urinary levels are increased in patients with 3-methylglutaconic aciduria type I