Learn More
Apexbio Technology LLC 3-Methylglutaric acid 626-51-7 500mg
SDP
Catalog No. 5000088933
Encompass_Preferred
3-Methylglutaric acid (CAS 626-51-7) is a metabolite derived from leucine degradation It accumulates as a result of defects in enzymes involved in leucine catabolism particularly in disorders such as 3-methylglutaconyl-CoA hydratase deficiency and 3-hydroxy-3-methylglutaryl-CoA lyase deficiency Elevated levels of 3-methylglutaric acid in biological samples serve as a biomarker for these inherited metabolic diseases The compound is utilized in research to study metabolic pathway disruptions enzyme function and the biochemical basis of organic acidurias linked to leucine metabolism
Catalog No. 50-000-88933 Supplier Apexbio Technology LLC Supplier No. B7868500
3-Methylglutaric acid (CAS 626-51-7) is a metabolite derived from leucine degradation It accumulates as a result of defects in enzymes involved in leucine catabolism particularly in disorders such as 3-methylglutaconyl-CoA hydratase deficiency and 3-hydroxy-3-methylglutaryl-CoA lyase deficiency Elevated levels of 3-methylglutaric acid in biological samples serve as a biomarker for these inherited metabolic diseases The compound is utilized in research to study metabolic pathway disruptions enzyme function and the biochemical basis of organic acidurias linked to leucine metabolism