Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylisoxazole-5-carbonyl chloride, 95%, Thermo Scientific™

Catalog No. CC27002DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC27002DA 1 g Amber Glass Bottle
CC27002CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. CC27002DA Supplier Thermo Scientific Chemicals Supplier No. CC27002DA
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 49783-72-4
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.54
MDL Number MFCD09817464
InChI Key VUBCNGCMOPTDED-UHFFFAOYSA-N
Synonym 3-methylisoxazole-5-carbonyl chloride, 3-methyl-5-isoxazolecarbonyl chloride, 5-isoxazolecarbonyl chloride, 3-methyl, aronis23810, 3-methylisoxazol-5-carbonyl chloride, 3-methyl-isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, 3-methyl-9ci
PubChem CID 12905099
IUPAC Name 3-methyl-1,2-oxazole-5-carbonyl chloride
SMILES CC1=NOC(=C1)C(Cl)=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 145.54
Percent Purity 95%
Chemical Name or Material 3-Methylisoxazole-5-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.