Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyloctane 99.0+%, TCI America™
SDP

Catalog No. M03095ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03095ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03095ML Supplier TCI America Supplier No. M03095ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2216-33-3
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00042906
InChI Key SEEOMASXHIJCDV-UHFFFAOYSA-N
PubChem CID 16664
IUPAC Name 3-methyloctane
SMILES CCCCCC(C)CC

Specifications

Color Colorless
Boiling Point 144°C
Quantity 5 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyloctane
EINECSNumber (2)-0009
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.