Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3-methyloxiran-2-yl)methanol | 872-38-8 | MFCD00033455 | 88.11 g/mol | C4H8O2 | 100 MG
SDP

Catalog No. 5000280789
Encompass_Preferred

(3-Methyloxiran-2-yl)methanol is a small-molecule drug intermediate used in organic and medicinal chemistry for the synthesis of various active compounds. The compound contains an epoxide alcohol functional group and is typically handled and stored under cold conditions to preserve stability.

  • Molecular formula: C4H8O2
  • Molecular weight: 88.11 g/mol
  • CAS number: 872-38-8
  • Purity: 95.0% (NMR)
  • Suitable for use as a drug intermediate in synthetic chemistry
  • Recommended storage: pure form -20°C for long term, 4°C for shorter term
  • Provided in small laboratory preparation sizes

Catalog No. 50-002-80789 Supplier Medchemexpress LLC Supplier No. HYW310318100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(3-Methyloxiran-2-yl)methanol is a small-molecule drug intermediate used in organic and medicinal chemistry for the synthesis of various active compounds. The compound contains an epoxide alcohol functional group and is typically handled and stored under cold conditions to preserve stability.

  • Molecular formula: C4H8O2
  • Molecular weight: 88.11 g/mol
  • CAS number: 872-38-8
  • Purity: 95.0% (NMR)
  • Suitable for use as a drug intermediate in synthetic chemistry
  • Recommended storage: pure form -20°C for long term, 4°C for shorter term
  • Provided in small laboratory preparation sizes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.