Learn More
Medchemexpress LLC (3-methyloxiran-2-yl)methanol | 872-38-8 | MFCD00033455 | 95.0% | 88.11 g·mol⁻1 | C4H8O2 | 50 MG
SDP
Catalog No. 5000282054
Encompass_Preferred
(3-Methyloxiran-2-yl)methanol is an epoxy-containing primary alcohol used as a synthetic building block and drug intermediate for small-molecule research. It has formula C4H8O2 (MW 88.11 g·mol⁻1) and CAS 872-38-8. Typical supplied purity is 95.0% (by NMR), and the product is offered in small research pack sizes.
Catalog No. 50-002-82054 Supplier Medchemexpress LLC Supplier No. HYW31031850MG
(3-Methyloxiran-2-yl)methanol is an epoxy-containing primary alcohol used as a synthetic building block and drug intermediate for small-molecule research. It has formula C4H8O2 (MW 88.11 g·mol⁻1) and CAS 872-38-8. Typical supplied purity is 95.0% (by NMR), and the product is offered in small research pack sizes.