Learn More
Medchemexpress LLC (3-methyloxiran-2-yl)methanol | 872-38-8 | MFCD00033455 | 95.0% | 88.11 g/mol | C4H8O2 | 250 MG
SDP
Catalog No. 5000281952
Encompass_Preferred
(3-Methyloxiran-2-yl)methanol is an epoxide-containing small-molecule intermediate used in medicinal chemistry and organic synthesis. It is employed as a building block for preparing active compound derivatives and for ring-opening transformations. Typical analytical values include formula C4H8O2, molecular weight 88.11 g/mol, CAS 872-38-8, and reported purity around 95%. Supplied in milligram pack sizes for research use only.
  • Epoxide functional group suitable for ring-opening reactions.
  • Low molecular weight facilitates handling and purification.
  • High reported purity for use as a synthetic intermediate.
  • Available in small pack sizes useful for research-scale synthesis.
  • Intended for research use only; not for clinical use.
Catalog No. 50-002-81952 Supplier Medchemexpress LLC Supplier No. HYW310318250MG
(3-Methyloxiran-2-yl)methanol is an epoxide-containing small-molecule intermediate used in medicinal chemistry and organic synthesis. It is employed as a building block for preparing active compound derivatives and for ring-opening transformations. Typical analytical values include formula C4H8O2, molecular weight 88.11 g/mol, CAS 872-38-8, and reported purity around 95%. Supplied in milligram pack sizes for research use only.
  • Epoxide functional group suitable for ring-opening reactions.
  • Low molecular weight facilitates handling and purification.
  • High reported purity for use as a synthetic intermediate.
  • Available in small pack sizes useful for research-scale synthesis.
  • Intended for research use only; not for clinical use.