Learn More
Medchemexpress LLC 3-Methylpyrazole | 1453-58-3 | 99.5% | C4H6N2 | 50 G
SDP
Supplier:  Medchemexpress LLC HY6605450G
Encompass_Preferred
3-Methylpyrazole is utilized as a nitrification inhibitor to control the nitrification process in soil. It has also been shown to inhibit the metabolism of methylazoxymethanol by rat liver microsomes.
  • Used as a nitrification inhibitor in soil.
  • Inhibits the metabolism of MAM by rat liver microsomes.
  • High purity of 99.54%.
Catalog No. 50-004-34288