Learn More
Selleck Chemical LLC 3-Methylpyrazole S5146-25mg
SDP
Catalog No. 503108439
Encompass_Preferred
3-Methylpyrazole (3-MP) is a weak or non-inhibitor of alcohol dehydrogenase
Catalog No. 50-310-8439 Supplier Selleck Chemical LLC Supplier No. S514625MG
3-Methylpyrazole (3-MP) is a weak or non-inhibitor of alcohol dehydrogenase