Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Methylpyridine

Catalog No. P4205325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P4205325ML 25 mL
P42053500ML 500 mL
P420531L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P4205325ML Supplier Sigma Aldrich Supplier No. P4205325ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Methylpyridine

Specifications

CAS 108-99-6
Melting Point -19°C (lit.)
Density 0.957 g/mL (at 25°C (literature))
Boiling Point 144°C (lit.)
Molecular Formula C6H7N
Refractive Index n20/D 1.504 (literature)
Linear Formula C6H7N
MDL Number MFCD00006402
Quantity 25 mL
Synonym 3-Picoline; 3-Methylpyridine
Molecular Weight (g/mol) 93.13
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.