Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpyrrole 98.0+%, TCI America™
SDP

Catalog No. M11441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11441G 1 g
M11445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11441G Supplier TCI America Supplier No. M11441G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-43-3
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00083419
InChI Key FEKWWZCCJDUWLY-UHFFFAOYSA-N
PubChem CID 12023
IUPAC Name 3-methyl-1H-pyrrole
SMILES CC1=CNC=C1

Specifications

Color Yellow
Boiling Point 144°C
Quantity 1 g
UN Number 1993
Formula Weight 81.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylpyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.