Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthio-1,2-propanediol 99%, Thermo Scientific™

Catalog No. AC206770250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206770250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC206770250 Supplier Thermo Scientific Chemicals Supplier No. 206770250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22551-26-4
Molecular Formula C4H10O2S
Molecular Weight (g/mol) 122.182
InChI Key FXIHXPKPLPKPJH-UHFFFAOYSA-N
Synonym 3-methylthio-1,2-propanediol, 3-methylthiopropane-1,2-diol, 3-methylsulfanyl propane-1,2-diol, s-methyl-thioglycerol, fxihxpkplpkpjh-uhfffaoysa, 1,2-propanediol,3-methylthio, 1,2-propanediol, 3-methylthio, 3-methylsulfanyl-1,2-propanediol #
PubChem CID 90802
IUPAC Name 3-methylsulfanylpropane-1,2-diol
SMILES CSCC(CO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.16
Infrared Spectrum Authentic
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
Linear Formula CH3SCH2CH(OH)CH2OH
Quantity 25 g
Formula Weight 122.18
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 3-Methylthio-1, 2-propanediol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.