Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthio-2-butanone 98.0+%, TCI America™
SDP

Catalog No. M111325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M111325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M111325ML Supplier TCI America Supplier No. M111325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53475-15-3
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD00008761
InChI Key HFVLNCDRAMUMCC-UHFFFAOYNA-N
Synonym 3-methylthio-2-butanone, 2-butanone, 3-methylthio, 3-methylmercapto-2-butanone, 3-methylsulfanyl butan-2-one, dl-3-methylthio-2-butanone, dl-3-methylthio butanone, acmc-1auk9, 2-butanone,3-methylthio, 3-methylsulfanyl-2-butanone
PubChem CID 103788
IUPAC Name 3-(methylsulfanyl)butan-2-one
SMILES CSC(C)C(C)=O

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
UN Number 1993
Formula Weight 118.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylthio-2-butanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.