Learn More
Medchemexpress LLC HY-101401 100mg Medchemexpress, 3-(Methylthio)propionic acid CAS:646-01-5 Purity:>98%
SDP
Catalog No. 502080630
Encompass_Preferred
Medchemexpress, HY-101401 100mg 3-(Methylthio)propionic acid CAS:646-01-5 3-(Methylthio)propionic acid is an intermediate in the methionine metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0630 Supplier Medchemexpress LLC Supplier No. HY101401100MG
Medchemexpress, HY-101401 100mg 3-(Methylthio)propionic acid CAS:646-01-5 3-(Methylthio)propionic acid is an intermediate in the methionine metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.