Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthiophene-2-carboxaldehyde 85.0+%, TCI America™
SDP

Catalog No. M0899100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0899100G 100 g
M089925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0899100G Supplier TCI America Supplier No. M0899100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5834-16-2
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005430
InChI Key BSQKBHXYEKVKMN-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarboxaldehyde, 3-methylthiophene-2-carboxaldehyde, 2-formyl-3-methylthiophene, 3-methylthiophene-2-aldehyde, 3-methyl-2-thiophenecarbaldehyde, 3-methyl-2-formylthiophene, 3-methyl-2-thiophene carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methyl, thiophenecarboxaldehyde, methyl, 3-methyl-thiophene-2-carbaldehyde
PubChem CID 79911
IUPAC Name 3-methylthiophene-2-carbaldehyde
SMILES CC1=C(SC=C1)C=O

Specifications

Boiling Point 113°C
Quantity 100 g
Formula Weight 126.17
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylthiophene-2-carboxaldehyde
EINECSNumber (5)-0074
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.