Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylthiophene-2-carboxaldehyde, tech. 85%

Catalog No. AAA1396114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1396114 25 g
AAA1396122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1396114 Supplier Thermo Scientific Chemicals Supplier No. A1396114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5834-16-2 | C6H6OS | 126.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5834-16-2
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005430
InChI Key BSQKBHXYEKVKMN-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarboxaldehyde, 3-methylthiophene-2-carboxaldehyde, 2-formyl-3-methylthiophene, 3-methylthiophene-2-aldehyde, 3-methyl-2-thiophenecarbaldehyde, 3-methyl-2-formylthiophene, 3-methyl-2-thiophene carboxaldehyde, 2-thiophenecarboxaldehyde, 3-methyl, thiophenecarboxaldehyde, methyl, 3-methyl-thiophene-2-carbaldehyde
PubChem CID 79911
IUPAC Name 3-methylthiophene-2-carbaldehyde
SMILES CC1=C(SC=C1)C=O

Specifications

Density 1.17
Boiling Point 215°C
Flash Point 82°C (179°F)
Refractive Index 1.586
Quantity 25 g
Beilstein 107874
Sensitivity Air Sensitive
Formula Weight 126.18
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Methylthiophene-2-carboxaldehyde
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 227-418-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.