Learn More
Selleck Chemical LLC 3-methylthiopropionate
SDP
Catalog No. 5000002943
Encompass_Preferred
3-(Methylthio)propionic acid (3-methylthiopropionate 4-Thiapentanoic acid 3-Methylsulfanylpropionic acid) is a thia fatty acid acid consisting of propionic acid with a methylthio substituent at the 3-position and an intermediate in mammalian methionine metabolism in vitro
Catalog No. 50-000-02943 Supplier Selleck Chemical LLC Supplier No. S634125UL
3-(Methylthio)propionic acid (3-methylthiopropionate 4-Thiapentanoic acid 3-Methylsulfanylpropionic acid) is a thia fatty acid acid consisting of propionic acid with a methylthio substituent at the 3-position and an intermediate in mammalian methionine metabolism in vitro